DWK Life Sciences Kimble™ 20mL Glass Screw-Thread Scintillation Vials
Manufactured from clear KG-33 borosilicate glass
Supplier:  DWK Life Sciences 7450020
 View more versions of this product
Catalog No. 36-099-770
Description
Description
Manufactured from clear KG-33 borosilicate glass
Variety of cap materials and liners available.
Order Info
Vials are packaged in cellular trays having alpha-numeric coordinates for fast and easy identification. Trays are shrink wrapped to preserve cleanliness. Each tray contains 100 vials, five trays per case. Caps come in polyethylene bags of 100.
Specifications
Specifications
Foil/Cork
Borosilicate Glass
Fast and easy identification
0.67 oz.
20 mL
Attached Screw Cap
ASTM E438
White
20 mL
Cellular Trays
Kimble
Urea
100 Cases (Vial), 100 Bag (Cap)
Scintillation Vial