Kimberly-Clark Professional™ WypAll™ X60 Wipers
Fast absorption and outstanding tear resistance
Manufacturer:  Kimberly-Clark Professional™ KC34015
 View more versions of this product
Catalog No. 29-633-990
Description
Description
Fast absorption and outstanding tear resistance
  • Made from Hydroknit™, an advanced wiper material manufactured using jets of water to bond soft, absorbent paper to extra-strong, nonwoven fabric
  • Reinforced for extra scrubbing and cleaning power
  • No adhesives or binders—soft enough for face and hands
Specifications
Specifications
White
General purpose cleaning and wiping of surfaces, equipment, and hands and faces
42.67 cm
Wypall™ X60 wipes
12.5 in.
16.8 in.
16.8 x 12.5 in. (42.67 x 31.75 cm)
16.8 in.
Hydroknit™ Paper
Yes
12.5 in.
31.75 cm
Documents