missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ WypAll™ Waterless Cleaning Wipes Refill
75 count refill bag for WypAll Waterless Cleaning Wipes Bucket. Ideal presaturated cleaner for on-the-go cleaning of hands, tools, parts and non-porous work surfaces. Natural solvents cut through grease, grime and oil while soothing ingredients aid in moisturizing the skin.
Manufacturer: Kimberly-Clark Professional™ KC91371
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
- 6 containers of 75 sheets per case; 450 sheets per case (9.5 in. x 12 in. each)
- Disposable industrial cleaning wipes are pre-saturated with an alcohol-free natural citrus cleaning agent for the waterless cleansing of skin and hard surfaces
- These wipers are tough on grease, but gentle enough to use on your hand; contains moisturizers to help keep skin hydrated
- One textured side for scrubbing dirt, grime, and grease (great for tool cleaning), one smooth side for wiping dirt off surfaces and hands
- Since these wipes are pre-moistened, you won't worry about spilling a liquid cleansing agent
Specifications
Bucket | |
Terpenes and Terpenoids (orange) 0 to 8%, Glycerin (Glycerol) 0 to 3%, non-hazardous ingredients 89 to 100% | |
Waterless Hand Wipe | |
Orange citrus |
450 Case | |
Green | |
Wipe | |
For cleaning of hands, tools, parts and non-porous work surfaces |