Kimberly-Clark Professional™ Sterile Evolution™ 4 Surgical Gowns
Fourth-generation fabric technology provides advanced protection, breathability and comfort
$351.33 - $351.33
Specifications
Certifications/Compliance | AORN |
---|---|
Closure Type | 2 Snaps |
Color | Blue |
Resistance | Fluid, flame and lint-resistant |
Sterility | Sterile |
Catalog Number | Mfr. No. | Material | Size | Sleeve Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Size | Sleeve Style | Price | Quantity | |||
19-810-266
|
Kimberly-Clark Professional™
KC90018 |
Non-reinforced Evolution 4 SMS | Large | Set-In-Sleeve |
Case of 36 Each for $351.33
|
|
Description
- Evolution 4 SMS fabric
- Adjustable neckline with yellow neck binding - hook and loop
- Resistant to fluid, flame and lint
- Meets NFPA Class I flame resistance
- Abrasion resistant
- Sterile
Set-in Sleeves
- Non-reinforced fabric
- Book fold
- Reinforced fabric
- Drop fold
Specifications
AORN | |
Blue | |
Sterile |
2 Snaps | |
Fluid, flame and lint-resistant |