Kimberly-Clark Professional™ Safeskin Purple Nitrile™ Sterile Exam Gloves
Kimberly-Clark™ Professional Safeskin Purple Nitrile™ Sterile Exam Gloves offer dexterity and tactile sensitivity while guarding against incidental splashes
Manufacturer:  Kimberly-Clark Professional™ KC55092
 View more versions of this product
Catalog No. 19-063-129
Description
Description
  • Made of a highly durable nitrile polymer
  • Powder free and not made with natural rubber latex (latex free)
  • Ambidextrous
  • Beaded cuffs prevent roll-down and allow for easy donning 
  • Excellent dexterity and tactile sensitivity, thanks to the textured fingertips
  • Fully recyclable through the RightCycle program (by Kimberly-Clark Professional), so lab gloves won’t end up in landfills
Specifications
Specifications
Yes
Beaded/Rolled
Individually packed
Powder-free
Sterile
24 cm
5 mil
Nitrile
Medium
Purple
9.5 in.