missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Purple Nitrile-XTRA™ Gloves

Kimberly-Clark™ Professional Purple Nitrile-XTRA™ Gloves are made of a highly durable nitrile polymer and are powder free.

 

Manufacturer:  Kimberly-Clark™ Professional KC50603

 View more versions of this product

Catalog No. 19-149864C




Description

Description

Availability Note: Due to temporary limited availability of these gloves, we have identified equivalent gloves that will meet your needs. For assistance in selecting the best option, please contact your sales specialist. We apologize for any inconvenience this may cause in the short term. You can direct support questions to safetyinquiries@thermofisher.com.

Contains no natural rubber latex proteins. Kimberly Clark™ Purple Nitrile-XTRA™ Gloves are ambidextrous, with textured fingertips for better grip.

  • Guard against incidental splashes
  • Not made with natural rubber latex (latex free)
  • Ambidextrous
  • Beaded cuff prevent roll-down and allow for easy donning
  • Excellent dexterity and tactile sensitivity, thanks to the textured fingertips
  • Cleared for use in chemotherapy.
  • Fully recyclable through the RightCycle program (by Kimberly-Clark Professional), so lab gloves won’t end up in landfills
Specifications

Specifications

Textured fingertips
Beaded/Rolled
Yes
Large
5.9mil
Powder-free
12 in.
Yes
Ambidextrous
Acrylonitrile Butadiene
21
ASTM D 6319, FDA, AQL 1.5
Purple
30.5cm
Documents
Promotions

Promotions