Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional KleenGuard™ A65 Flame-Resistant Lab Coats

Designed for use in areas where exposure to sparks or flame is possible

$222.42 - $245.77

Specifications

Certifications/Compliance NFPA 701, CAN CGSB-155.20-2000, NFPA 99
Closure Type Hook and loop
Color Blue
For Use With (Application) Ideal for use in laboratories, use in areas where exposure to sparks or flame is possible
Resistance Flame, tear
View More Specs
Products
Catalog Number Mfr. No. Material Size Price Quantity    

19-047-720

 
kimberly-clark™ professional
KC12810
Cotton Case of 25 Each for $241.92

19-047-724

 
kimberly-clark™ professional
KC12814
Cotton Case of 25 Each for $233.43

19-047-723

 
kimberly-clark™ professional
KC12813
Cotton Case of 25 Each for $222.42

19-047-725

 
kimberly-clark™ professional
KC12815
Cotton Case of 25 Each for $245.77
Description & Specifications

Specifications

Certifications/Compliance NFPA 701, CAN CGSB-155.20-2000, NFPA 99
Closure Type Hook and loop
Color Blue
For Use With (Application) Ideal for use in laboratories, use in areas where exposure to sparks or flame is possible
Resistance Flame, tear

Ideally suited for use in laboratories.

  • Comfortable, breathable and durable fabric resists tearing
  • Hook and loop closures
  • No pocket
  • May also be used to protect primary fire protection clothing

Meets NFPA 701 FLAME PROPAGATION TEST, 1989 Edition. Complies with the flame-resistance requirements cited by paragraph 6.1 of CAN/CGSB-155.20-2000. NFPA 99 compliant antistatic material.