Kimberly-Clark Professional™ KleenGuard™ A65 Flame-Resistant Coveralls
Soft, lightweight and breathable
Manufacturer:  Kimberly-Clark Professional™ KC45312
 View more versions of this product
Catalog No. 29-633-944
Description
Description
Designed to be worn in areas with potential exposure to sparks, these coveralls are made of a comfortable, breathable and durable fabric treated with a flame-resistant finish.
  • Design meets ANSI™/ISEA™ 101-1996 sizing standard
  • Ideal for welding, oil refining, utilities, grinding and general maintenance applications
  • Zipper front
Compliance
While not intended as primary fire protection apparel, the fabric is flame-resistant and compliant with NFPA™ 701-1989 Flame Propagation test, meaning it will not melt or propagate a flame.
Specifications
Specifications
Open
Fabric
ANSI ISEA 101-1996, CAN CGSB-155.20-2000, NFPA 701, NFPA 99
Open
Welding, Oil refining, utilities, grinding
88.26 cm
38 to 40 in.
40 in.
76.2 cm
Front zipper
Zipper Front
Blue
Durable, spunlace, non-woven Fabric treated with flame-resistant finish
34.75 in.
Medium
30 in.
96.52 to 101.6 cm
101.6 cm