missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ KleenGuard™ A65 Flame-Resistant Coveralls

Soft, lightweight and breathable

$304.77 - $391.94

Specifications

Certifications/Compliance ANSI ISEA 101-1996, CAN CGSB-155.20-2000, NFPA 701, NFPA 99
Closure Type Front zipper
Color Blue
For Use With (Application) Welding, Oil refining, utilities, grinding
View More Specs

Products 16
Catalog Number Mfr. No. Ankle Style Cuff Style Style Size Price Quantity  
Catalog Number Mfr. No. Ankle Style Cuff Style Style Size Price Quantity  
29-633-944
View Documents Promotion Details
Kimberly-Clark Professional™
KC45312
Open Open Zipper Front Medium
Case of 25 Each for $318.06
 
29-633-945
View Documents Promotion Details
Kimberly-Clark Professional™
KC45313
Open Open Zipper Front Large
Case of 25 Each for $318.06
 
29-633-948
View Documents Promotion Details
Kimberly-Clark Professional™
KC45316
Open Open Zipper Front 3X-Large
Case of 21 Each for $318.92
 
29-633-947
View Documents Promotion Details
Kimberly-Clark Professional™
KC45315
Open Open Zipper Front 2X-Large
Case of 25 Each for $304.77
 
29-633-950
View Documents Promotion Details
Kimberly-Clark Professional™
KC45323
Elastic Elastic Zipper Front, Attached Hood Large
Case of 25 Each for $369.46
 
29-633-951
View Documents Promotion Details
Kimberly-Clark Professional™
KC45324
Elastic Elastic Zipper Front, Attached Hood X-Large
Case of 25 Each for $369.46
 
29-633-946
View Documents Promotion Details
Kimberly-Clark Professional™
KC45314
Open Open Zipper Front X-Large
Case of 25 Each for $322.49
 
29-633-952
View Documents Promotion Details
Kimberly-Clark Professional™
KC45325
Elastic Elastic Zipper Front, Attached Hood 2X-Large
Case of 25 Each for $357.13
 
29-633-953
View Documents Promotion Details
Kimberly-Clark Professional™
KC45326
Elastic Elastic Zipper Front, Attached Hood 3X-Large
Case of 21 Each for $346.00
 
29-635-2215
View Documents Promotion Details
Kimberly-Clark Professional™
KC45352
Elastic Elastic Zipper Front, Attached Hood and Boots Medium
Case of 25 Each for $390.19
 
29-635-2220
View Documents Promotion Details
Kimberly-Clark Professional™
KC45357
Elastic Elastic Zipper Front, Attached Hood and Boots 4X-Large
Case of 21 Each for $391.94
 
29-635-2217
View Documents Promotion Details
Kimberly-Clark Professional™
KC45354
Elastic Elastic Zipper Front, Attached Hood and Boots X-Large
Case of 25 Each for $390.19
 
29-635-2218
View Documents Promotion Details
Kimberly-Clark Professional™
KC45355
Elastic Elastic Zipper Front, Attached Hood and Boots 2X-Large
Case of 25 Each for $390.19
 
29-635-2213
View Documents Promotion Details
Kimberly-Clark Professional™
KC45317
Open Open Zipper Front 4X-Large
Case of 21 Each for $318.92
 
19-457-057G
View Documents Promotion Details
Kimberly-Clark Professional™
KC42000
Open Open Zipper Front 5X-Large
Case of 21 Each for $324.70
 
29-635-2216
View Documents Promotion Details
Kimberly-Clark Professional™
KC45353
Elastic Elastic Zipper Front, Attached Hood and Boots Large
Case of 25 Each for $390.19
 
Description

Description

Designed to be worn in areas with potential exposure to sparks, these coveralls are made of a comfortable, breathable and durable fabric treated with a flame-resistant finish.

  • Design meets ANSI™/ISEA™ 101-1996 sizing standard
  • Ideal for welding, oil refining, utilities, grinding and general maintenance applications
  • Zipper front

Compliance

While not intended as primary fire protection apparel, the fabric is flame-resistant and compliant with NFPA™ 701-1989 Flame Propagation test, meaning it will not melt or propagate a flame.

Specifications

Specifications

ANSI ISEA 101-1996, CAN CGSB-155.20-2000, NFPA 701, NFPA 99
Front zipper
Blue
Welding, Oil refining, utilities, grinding
SDS
Product Certifications