Kimberly-Clark Professional™ KleenGuard™ A65 Flame-Resistant Coveralls
Soft, lightweight and breathable
Manufacturer:  Kimberly-Clark Professional™ KC45357
 View more versions of this product
Catalog No. 29-635-2220
Description
Description
Designed to be worn in areas with potential exposure to sparks, these coveralls are made of a comfortable, breathable and durable fabric treated with a flame-resistant finish.
  • Design meets ANSI™/ISEA™ 101-1996 sizing standard
  • Ideal for welding, oil refining, utilities, grinding and general maintenance applications
  • Zipper front
Compliance
While not intended as primary fire protection apparel, the fabric is flame-resistant and compliant with NFPA™ 701-1989 Flame Propagation test, meaning it will not melt or propagate a flame.
Specifications
Specifications
Elastic
Polypropylene
Blue
37 in.
4X-Large
32 in.
81.28 cm
Front zipper
Zipper Front, Attached Hood and Boots
Elastic
14.56 cm
58 to 60 in.
56 in.
142.24 cm