Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Lab Coats

Lab coats offer bloodborne pathogen and chemical splash protection for applications including forensics and animal research

$200.48 - $200.48

Specifications

Certifications/Compliance ASTM F1670, 1671
Closure Type Snap Front
Color Blue
Construction 3-ply SMS
Material Microporous film, Spunbond Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Size (English) Chest Size Price Quantity    

29-633-837

 
kimberly-clark™ professional
KC45514
29.75 in. X-Large Case of 25 Each for $200.48
Description & Specifications

Specifications

Certifications/Compliance ASTM F1670, 1671
Closure Type Snap Front
Color Blue
Construction 3-ply SMS
Material Microporous film, Spunbond Polypropylene
No. of Pockets 2.0
Disposable Yes
For Use With (Application) Suited for forensics and animal research
Seam Type Serged

  • Patented three-layer fabric construction
  • Middle layer is a breathable microporous film that offers resistance against many liquids and dry particulates while also allowing heat and sweat vapor to escape
  • Outer layers are a cloth-like yet tough and abrasion-resistant spunbond polypropylene
  • Snap-front, with chest and hip pocket

Fabric passes ASTM™ F1670/F1671 testing for penetration of blood, bodily fluids, and bloodborne pathogens.