Kimberly-Clark Professional™ KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Lab Coats
Lab coats offer bloodborne pathogen and chemical splash protection for applications including forensics and animal research
Manufacturer:  Kimberly-Clark Professional™ KC45514
 View more versions of this product
Catalog No. 29-633-837
Description
Description
  • Patented three-layer fabric construction
  • Middle layer is a breathable microporous film that offers resistance against many liquids and dry particulates while also allowing heat and sweat vapor to escape
  • Outer layers are a cloth-like yet tough and abrasion-resistant spunbond polypropylene
  • Snap-front, with chest and hip pocket
Compliance
Fabric passes ASTM™ F1670/F1671 testing for penetration of blood, bodily fluids, and bloodborne pathogens.
Specifications
Specifications
ASTM F1670, 1671
Blue
2
Suited for forensics and animal research
Serged
3-ply SMS
Snap
Microporous Film,Spunbond Polypropylene
X-Large
Yes
29.75 in.
Documents