missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Coveralls
Provide exceptional protection against nonhazardous liquids, dry particulates, and bloodborne pathogens, even under pressure
Manufacturer: Kimberly-Clark Professional™ KC45026
Description
Breathable fabric is strong, comfortable, and abrasion-resistant, with a three-layer construction designed to allow heat and sweat vapor to escape.
- Patented Reflex™ Design is 7.5 times less likely to rip out than ANSI™ minimum standards
- Feature 12% more chest room and 6% longer body length
- Provide excellent front and strikethrough protection with a seamless front and concealed 28 in. (71cm) zipper
- Used in many industries and applications, including emergency medical, liquid handling, forensics, nuclear facilities and more
- Denim blue
Compliance
Meet ASTM F1670/1671 specifications for blood, bodily fluids and bloodborne pathogens.
Specifications
Elastic | |
Fabric | |
ASTM F1670, 1671 | |
Elastic | |
Animal research, emergency medical response, crime scene cleanup, embalming/forensics, tank cleaning, Fiberglass manufacturing, liquid handling, food processing | |
3X-Large |
Front zipper | |
Elastic | |
Denim Blue | |
Hooded, seamless zipper front with storm flap, elastic wrist and ankle, elastic across the back; 3-ply Fabric construction middle layer of Microporous film | |
Hooded |