Kimberly-Clark Professional™ KleenGuard™ A40 Liquid and Particle Protection Lab Coats
Provides better liquid and particulate barrier
Manufacturer:  Kimberly-Clark Professional™ KC44444
 View more versions of this product
Catalog No. 19-120-2403D
Description
Description
Suited for use in fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance and general manufacturing
  • Snap front closure
  • Elastic around the wrists
  • Available with or without pockets
  • Breathable, microporous film laminate
  • Low in lint
  • Traditional collar
  • Passes NFPA 99 criteria for antistatic materials
Compliance
Meets ASTM F1670 testing for penetration of blood and bodily fluids
Specifications
Specifications
Microporous Film Laminate
NFPA 99, ASTM F1670, ANSI ISEA 101-1996
White
Suited for Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance and general manufacturing
X-Large
Traditional
Snaps
Elastic
Polypropylene