Kimberly-Clark Professional™ KleenGuard™ A40 Liquid and Particle Protection Lab Coats
Provides better liquid and particulate barrier
Manufacturer: Kimberly-Clark Professional™ KC44445
Description
Suited for use in fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance and general manufacturing
- Snap front closure
- Elastic around the wrists
- Available with or without pockets
- Breathable, microporous film laminate
- Low in lint
- Traditional collar
- Passes NFPA 99 criteria for antistatic materials
Compliance
Meets ASTM F1670 testing for penetration of blood and bodily fluids
Specifications
Microporous Film Laminate | |
NFPA 99, ASTM F1670, ANSI ISEA 101-1996 | |
White | |
Suited for Fiberglass manufacturing, tank cleaning, utilities, liquid handling, pressure washing, maintenance and general manufacturing | |
2X-Large |
Traditional | |
Snaps | |
Elastic | |
Polypropylene |