Kimberly-Clark Professional™ KleenGuard™ A40 Liquid and Particle Protection Coveralls
Excellent protection from nonhazardous liquids
Manufacturer: Kimberly-Clark Professional™ KC44334
Description
Excellent protection from nonhazardous liquids
Microporous, nonwoven film laminate offers an improved liquid barrier over SMS apparel.
- Low linting
- White
- Zipper front
Compliance
Meets ANSI™/ISEA™ 101-1996; ASTM™ F1670 testing for penetration of blood and bodily fluids; NFPA 99 Health Care Facilities criteria for antistatic materials.
Specifications
Elastic | |
Polypropylene | |
White | |
Zipper Front, Elastic Wrists, Ankles, Hood and Boots; Non-woven Microporous film laminate barrier over SMS | |
Fiberglass manufacturing, liquid handling, utilities, pressure washing, tank cleaning, maintenance and general manufacturing | |
X-Large |
Front zipper | |
ANSI/ISEA 101-1996, ASTM F1670, NFPA 99 | |
Elastic | |
Yes | |
Hooded |