Kimberly-Clark™ Professional KleenGuard™ A40 Liquid and Particle Protection Boot and Shoe Covers
Manufacturer:  Kimberly-Clark™ Professional 44495DUP
Catalog No. 29-635-9215
Description
Description
  • Constructed of breathable microporous film laminate low-linting fabric
  • Universal size has elastic down sole; other sizes feature vinyl soles
  • Packed for use in cleanroom
Compliance
Meets NFPA™ 99 standards for Antistatic Material and ASTM™ F1670 testing for penetration of blood and bodily fluids.