Kimberly-Clark™ Professional KleenGuard™ A30 Breathable Splash Particle Protection Coveralls with Elastic Back, Wrists and Ankles
Manufacturer:  Kimberly-Clark™ Professional KC46102DUP
Catalog No. 29-638-1593
Description
Description
MICROFORCE™ Barrier SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids. Suitable for many applications including pharmaceutical research, food processing and more.
  • Excellent protection from nonhazardous particles and liquids
  • Strong and abrasion resistant; suitable for multiple wearings
  • Seamless front to provide more protection in the primary exposure areas
  • Zipper front with 1 in. (2.5cm) flap and elastic back
  • Color: White
  • With elastic back, wrists and ankles
Compliance
Passes NFPA 99 for antistatic materials.