missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A30 Breathable Splash Particle Protection Coveralls with Elastic Back, Wrists and Ankles and Attached Hood
Combine durability and breathability to provide comfortable splash and particle protection
Manufacturer: Kimberly-Clark Professional™ KC46115
Description
MICROFORCE™ Barrier SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids. Suitable for many applications including pharmaceutical research, food processing and more.
- Excellent protection from nonhazardous particles and liquids
- Strong and abrasion resistant; suitable for multiple wearings
- Seamless front to provide more protection in the primary exposure areas
- Zipper front with 1 in. (2.5cm) flap and elastic back
- Color: White
- With elastic back, wrists and ankles and attached hood
Compliance
Passes NFPA 99 for antistatic materials.
Specifications
Elastic | |
Elastic | |
Elastic |
Polypropylene | |
White | |
2X-Large |