Kimberly-Clark™ Professional KleenGuard™ A30 Breathable Splash Particle Protection Coveralls with Elastic Back, Wrists and Ankles
Supplier: Kimberly-Clark™ Professional 46106
Description
MICROFORCE™ Barrier SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids. Suitable for many applications including pharmaceutical research, food processing and more.
- Excellent protection from nonhazardous particles and liquids
- Strong and abrasion resistant; suitable for multiple wearings
- Seamless front to provide more protection in the primary exposure areas
- Zipper front with 1 in. (2.5cm) flap and elastic back
- Color: White
- With elastic back, wrists and ankles
Compliance
Passes NFPA 99 for antistatic materials.