missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A30 Breathable Splash Particle Protection Coveralls with Elastic Back
Combine durability and breathability to provide comfortable splash and particle protection
Supplier: Kimberly-Clark Professional™ 46007
Description
MICROFORCE™ Barrier SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids. Suitable for many applications including pharmaceutical research, food processing and more.
- Excellent protection from nonhazardous particles and liquids
- Strong and abrasion resistant; suitable for multiple wearings
- Seamless front to provide more protection in the primary exposure areas
- Zipper front with 1 in. (2.5cm) flap and elastic back
- Color: White
- With elastic back
Compliance
Passes NFPA 99 for antistatic materials.
Specifications
4X-Large | |
White | |
Open |
Polypropylene | |
Open | |
Elastic |