Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional KleenGuard™ A20 Lab Coat with Knit Cuffs

Breathable particle protection with comfortable snug fit

$62.00 - $179.81

Specifications

Certifications/Compliance NFPA 99
Collar Type High mandarin
Closure Type Snap
Color Denim Blue
Material Microforce™ barrier SMS Fabric
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    

19-999-974

 
kimberly-clark™ professional
KC23871
38 in. 97cm Small Case of 15 Each for $179.81

19-999-975

 
kimberly-clark™ professional
KC23872
40 in. 102cm Medium Case of 15 Each for $179.81

19-999-976

 
kimberly-clark™ professional
KC23873
42 in. 107cm Large Case of 15 Each for $179.81

19-999-977

 
kimberly-clark™ professional
KC23874
44 in. 112cm X-Large Case of 15 Each for $179.81

19-020-2500

 
kimberly-clark™ professional
KC23875
46 in. 117cm 2X-Large Case of 15 Each for $62.00
Description & Specifications

Specifications

Certifications/Compliance NFPA 99
Collar Type High mandarin
Closure Type Snap
Color Denim Blue
Material Microforce™ barrier SMS Fabric
No. of Pockets 2
For Use With (Application) Suited for laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments

Jackets feature snug-fitting polyester-knit cuffs on raglan sleeves.

  • With high mandarin collar, one chest pocket and one hip pocket
  • Plastic snap-front closure
  • Denim blue

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.