Kimberly-Clark Professional™ KleenGuard™ A20 Breathable Particle Protection Shirt
Kimberly-Clark™ Professional KleenGuard™ A20 Breathable Particle Protection Shirt protects against hazardous and non-hazardous dust
Manufacturer:  Kimberly-Clark Professional™ KC36214
 View more versions of this product
Catalog No. 29-636-2484
Description
Description
  • Designed to help protect against dirt, grime and dust 
  • Five layers of stacked barrier protection (SMMMS) against hazardous particles
  • Hip-length shirt
  • Comes in a variety of sizes
  • Serged seams
  • Five snap closures
  • Open wrists
  • Mandarin collar
  • Popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments
  • Made with breathable Microforce barrier fabric
Specifications
Specifications
Passes NFPA 99 criteria for antistatic materials
White
X-Large
Snaps
Fabric