Kimberly-Clark Professional™ KleenGuard™ A20 Breathable Particle Protection Denim Blue Coveralls
Strong, breathable, abrasion-resistant fabric keep workers cool and comfortable while protecting against fibers, fine dusts, and low surface tension liquid splashes
Manufacturer:  Kimberly-Clark Professional™ KC58536
 View more versions of this product
Catalog No. 19-057-400E
Description
Description
Patented Reflex™ design is 7.5 times less likely to rip out than ANSI™ minimums — provides 12% more chest room and 6% longer body length.
  • Ideal for use in dusty environments, as well as for painting, manufacture spraying, and agricultural spraying
  • Stylish denim blue color masks dirt and grease
  • Air and moisture vapor can pass through to aid wearer comfort
  • Triple-stitched serged seams
  • Can withstand multiple wearing
  • Zipper front
  • Open wrists and ankles
Specifications
Specifications
Open
SMS
Denim Blue
Microforce™ Barrier SMS Fabric
Serged
Front zipper
ANSI, NFPA 99
Open
Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
3X-Large