missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kimberly-Clark Professional™ KleenGuard™ A10 Light-Duty Coveralls
The right combination of price and protection for nonhazardous environments
Supplier: Kimberly-Clark Professional™ KC10610
Description
- Lightweight, breathable fabric keeps workers cool and comfortable
- Zipper front
- Elastic wrists
- Also available with attached hood and boots
- White
Compliance
Meets ANSI™/ISEA™ 101-1996 standards.
Specifications
Light-duty Coveralls | |
Polypropylene | |
Attached Boots | |
Hooded | |
Serged | |
With attached hood and boots; Breathable SMS Polypropylene |
3X-Large | |
White | |
Elastic | |
Front Zipper | |
ANSI/ISEA 101-1996 | |
Food processing, general maintenance, manufacturing |