missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Kimtech Pure™ W4 Wipers

For ISO Class 4 and higher applications

Manufacturer:  Kimberly-Clark Professional™ KC06070

 View more versions of this product

Catalog No. 19-130-1574


Alert:

Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.


Description

Description

  • Made of 100% polypropylene that resists acids, bases and solvents
  • Thermally bonded—no glues or binders
  • Straight, clean, knife-cut edges minimize lint

Order Info

Dry wipers flat packed and double bagged with a case liner.

Specifications

Specifications

11 in.
White
For ISO Class 4 and higher cleanroom applications
28 cm
W4 Presaturated wipes
40 sheets/pack
Non-sterile
28 cm
11 x 9 in. (28 x 23 cm)
11 in.
Polypropylene
Straight clean knife-cut
70% Isopropyl Alcohol, 30% Deionised Water
SDS
Documents