missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Kimtech Pure™ W4 Wipers

For ISO Class 4 and higher applications

Manufacturer:  Kimberly-Clark Professional™ KC33390

 View more versions of this product

Catalog No. 19-130-1641


Alert:

Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.


Description

Description

  • Made of 100% polypropylene that resists acids, bases and solvents
  • Thermally bonded—no glues or binders
  • Straight, clean, knife-cut edges minimize lint

Order Info

Dry wipers flat packed and double bagged with a case liner.

Specifications

Specifications

9 in.
FDA
9 x 9 in. (23 x 23 cm)
9 in.
Polypropylene
Straight clean knife-cut
Non-sterile
23 cm
White
For ISO Class 4 and higher cleanroom applications
23 cm
W4 Dry wipes
100 sheets/pack
SDS
Documents
Product Certifications