Kimberly-Clark™ Professional Kimtech Pure™ M3 Facemasks
Pleat-style mask with soft ties or ear loops
$17.25 - $201.87
Specifications
Color White
Material SMS, Polypropylene
Size Regular
Latex Free Yes
Pore Size 0.1 (PFE), 3 (BFE)μ
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Sterile Includes Price Quantity  
Catalog Number Mfr. No. Sterile Includes Price Quantity  
18-999-4775 Kimberly-Clark™ Professional
KC62466
No Soft ties Pack of 50 Each for $17.50
Case of 10 Pack for $159.80
18-999-4788 Kimberly-Clark™ Professional
KC62494
Yes Soft ties Case of 200 Each for $201.87
18-999-4827 Kimberly-Clark™ Professional
KC62465
No Knitted earloops Pack of 50 Each for $17.25
Case of 10 Pack for $157.50
Specifications
Description
  • Inner facing made of white BiCoSof™
  • Nose piece made from fully enclosed, soft, malleable aluminium
  • Outer facing made from clear polyethylene apertured film
  • Loops (62466): white, tubular knitted polyester
Order Info
Certificate of Irradiation available for sterile mask.
Description & Specifications
Specifications
White
Regular
0.1 (PFE), 3 (BFE)μ
SMS, Polypropylene
Yes
Pleated