Kimberly-Clark™ Professional Kimtech Pure™ G5 Nitrile Gloves
Manufacturer-suggested ISO Class 5
Manufacturer:  Kimberly-Clark™ Professional KC56860
 View more versions of this product
Catalog No. 11-390-13B
Specifications Description & Specifications
Specifications
Nitrile
10 in.
5.1mil
Powder-free
Bisque
Medium
Beaded/Rolled
Blue
25.4cm
ISO Class 5
Ambidextrous