missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Kimtech Pure™ G3 Nonsterile Latex Hand-Specific Gloves

Recommended for ISO Class 4 or higher cleanroom environments

Manufacturer:  Kimberly-Clark™ Professional KC56828

 View more versions of this product

Catalog No. 11-390-10B


 

Alert:

The products listed on this page contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Specifications Description & Specifications

Specifications

12 x 3.26 in. (30.5 x 8.3cm)
Hand-specific
7.5
8.3cm
Powder-free
Sterile
30.5cm
Beaded/Rolled
Rubber
3.26 in.
7.9mil
Natural
12 in.