Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Kimtech Pure™ G3 Nonsterile Latex Hand-Specific Gloves


Recommended for ISO Class 4 or higher cleanroom environments

Manufacturer: kimberly-clark™ professional  KC56834

 View more versions of this product

 Alert

The products listed on this page contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Catalog No. 11-390-8H

  • / Pack of 50 Pair
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Cuff Style Beaded cuff
Puncture Rating AQL level of 1.5 for pinholes
Palm Type Textured
Glove Cuff Style Beaded
Material Rubber
Size 10
Thickness 8.7mil
Hand Ambidextrous
Color Natural
Length (Metric) 30.5cm
Sterile No
Length (English) 12 in.