Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Kimtech Pure™ G3 Nonsterile Latex Hand-Specific Gloves


Recommended for ISO Class 4 or higher cleanroom environments

Manufacturer: kimberly-clark™ professional  KC56840

 View more versions of this product

 Alert

The products listed on this page contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Catalog No. 11-390-8F

  • / Pack of 50 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Thickness (English) 8.7mil
Sterility Non-sterile
Glove Cuff Style Beaded/Rolled
Length (English) 12 in.
Color Natural
Length (Metric) 30.5cm
Size 8.5
Hand Ambidextrous
Material Rubber