Please login to your online account to display your discounted pricing
Kimberly-Clark™ Professional Kimtech Pure™ G3 Nonsterile Latex Hand-Specific Gloves
Recommended for ISO Class 4 or higher cleanroom environments
Manufacturer: kimberly-clark™ professional  KC56837
 View more versions of this product
 Alert
The products listed on this page contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 11-390-8C
  • / Pack of 50 Each
  • / Case of 4 Pack
Description & Specifications
Specifications
Glove Cuff Style Beaded/Rolled
Sterility Non-sterile
Thickness (English) 8.7mil
Length (English) 12 in.
Length (Metric) 30.5cm
Size 7
Color Natural
Material Rubber
Hand Ambidextrous