missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Kimtech Pure™ G3 and G5 STERLING™ Nitrile Gloves

Combine the protection of nitrile with the sensitivity of latex

$291.72 - $311.20

Specifications

Color Gray
Length (English) 12 in.
Length (Metric) 30.48cm
Material Nitrile
Sterility Non-sterile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Classification Size Price Quantity  
Catalog Number Mfr. No. Classification Size Price Quantity  
17-986-007 Kimberly-Clark™ Professional
KC98184
ISO Class 5 X-Small Case of 1500 Each for $291.72
17-986-008 Kimberly-Clark™ Professional
KC98185
ISO Class 5 Small Case of 1500 Each for $291.72
17-986-010 Kimberly-Clark™ Professional
KC98187
ISO Class 5 Large Case of 1500 Each for $291.72
17-986-011 Kimberly-Clark™ Professional
KC98188
ISO Class 5 2X-Large Case of 1500 Each for $291.72
17-986-009 Kimberly-Clark™ Professional
KC98186
ISO Class 5 Medium Case of 1500 Each for $291.72
17-986-012 Kimberly-Clark™ Professional
KC98189
ISO Class 5 X-Large Case of 1500 Each for $291.72
17-986-001 Kimberly-Clark™ Professional
KC99234
ISO Class 3 X-Small Case of 1500 Each for $311.20
17-986-003 Kimberly-Clark™ Professional
KC99236
ISO Class 3 Medium Case of 1500 Each for $311.20
17-986-002 Kimberly-Clark™ Professional
KC99235
ISO Class 3 Small Case of 1500 Each for $311.20
17-986-004 Kimberly-Clark™ Professional
KC99237
ISO Class 3 Large Case of 1500 Each for $311.20
17-986-005 Kimberly-Clark™ Professional
KC99238
ISO Class 3 2X-Large Case of 1500 Each for $311.20
17-986-006 Kimberly-Clark™ Professional
KC99239
ISO Class 3 X-Large Case of 1500 Each for $311.20
Specifications

Description

Compared to traditional latex and nitrile gloves, the new KIMTECH PURE G3 and G5 STERLING Nitrile Gloves offer better performance, better value, and are better for the environment. The process also allows a reduction in glove thickness, so less force is required to stretch the material. The result is a latex-free glove with the dexterity and sensitivity of latex.

  • G3 is recommended for ISO Class 3 or higher cleanrooms; G5, for ISO Class 5 or higher cleanrooms
  • Natural rubber latex-free, reducing the potential for TYPE 1 glove-associated reactions
  • Static dissipative in use
  • Certificate of Analysis (by lot) available online
  • Textured fingertips
  • Beaded cuff; comfortable fit
  • Nonsterile
  • Powder-free
  • Excellent dexterity and tactile sensitivity
  • Ambidextrous with high tack/grip
  • Thickness: 4.0mil
  • Length: 12 in. (30.5cm)
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail

Applications:

Semiconductor industry, medical device, flat panel display, pharmaceutical, disk drive manufacturing, and electronics.

Order Info

Double poly bagged with poly case liner.

Description & Specifications

Specifications

Gray
30.48cm
Non-sterile
Beaded/Rolled
Powder-free
12 in.
Nitrile
Textured
Ambidextrous
4.0mil