Kimberly-Clark Professional™ Kimtech™ A5 Sterile Hood with Ties
Sterile cleanroom hoods with secure ties featuring innovative CLEAN-DON™ technology for simpler aseptic donning. Suitable for EU GMP ISO 5 Grade A sterile cleanrooms, the hoods are individually vacuum-packed and made from light and breathable fabric.
Manufacturer: Kimberly-Clark Professional™ KC88807
Description
- Suitable for use in EU GMP ISO 5 Class A sterile cleanroom environments and laboratories, and provided with supporting technical information to demonstrate regulatory compliance.
- Advanced CLEAN-DON™ technology for fast, simple and compliant aseptic donning that combines two donning steps into one.
- The comfortable hoods are provided in a universal size and are made from low-lint breathable SMS fabric with a cloth-like feel.
- Contamination protection is ensured through high quality sterility assured material with high-strength bound and triple-stitched seams, and secure ties.
- Individually vacuum-packed and efficiently double-bagged to save space.
- Assured Compliance:
- PPE Cat III according to Regulation (EU) 2016/425
- Type 6 (PB) certified
- Certificate of Conformance by lot available online
- Certificate of Sterility by lot available online.
Order Info
Gamma sterilized and individually bagged. Certificate of Irradiation available.
Specifications
100/Cs. | |
SMS | |
Without Ties | |
Universal |
White | |
ISO 5 to 8 cleanroom environments, compounding, mixing, filling, cleaning | |
SMS | |
Sterile |