Kimberly-Clark Professional™ KleenGuard™ Visio™ Safety Glasses
For eye protection that’s both economical and reliable, you can trust Visio Safety Glasses from KleenGuard.
Manufacturer:  Kimberly-Clark Professional™ 14470
 View more versions of this product
Catalog No. 19-059327
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
Description
  • One-piece polycarbonate front provides 99.9% UV protection
  • Sleek, lightweight, and comfortable
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
Meets ANSI Z87.1-2010 standards.
Uncoated
Safety glasses
Silver
Clear