Kimberly-Clark Professional™ KleenGuard™ G60 Level 2 Nitrile Coated Cut Resistant Gloves
Choose KleenGuard G60 Level 2 Nitrile Coated Cut Resistant Gloves when you need dexterity, grip and cut resistance for risk of minor cuts and abrasions.
Manufacturer:  Kimberly-Clark Professional™ KC98230
 View more versions of this product
Catalog No. 19-065-791
Description
Description
Used for applications like metal fabrication, glass handling, stamping, automotive and aerospace assembly.
  • Stretch Kevlar™ liner provides cut resistance and a comfortable fit
  • Dexterity for improved productivity
  • Nitrile coating enhances grip in light oil applications
  • Dark coating hides dirt for longer wear
Compliance
ANSI™ Certified Level 2
Specifications
Specifications
Level 2
Nitrile
Small
ANSI Level 2/EN388 Level 2 cut resistance
Metal fabrication, Glass handling, stamping, automotive and aerospace assembly
Stretch Dupont™ Kevlar™ aramid fiber
Blue,Yellow