missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional Jackson Saftey™ G60 Nitrile Foam Cut-Resistant Gloves

Useful for working environments that pose a risk of minor cuts and abrasions

$399.23 - $399.23

Specifications

Color Blue,Yellow
Cut Resistance Rating Level 2
Material Nitrile
For Use With (Application) Metal fabrication, Glass handling, stamping, automotive and aerospace assembly
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-065-791 Kimberly-Clark™ Professional
KC98230
Small Case of 60 Pair for $399.23
19-065-792 Kimberly-Clark™ Professional
KC98231
Medium Case of 60 Pair for $399.23
19-065-793 Kimberly-Clark™ Professional
KC98232
Large Case of 60 Pair for $399.23
19-065-794 Kimberly-Clark™ Professional
KC98233
X-Large Case of 60 Pair for $399.23
Specifications

Description

Used for applications like metal fabrication, glass handling, stamping, automotive and aerospace assembly.

  • Stretch Kevlar™ liner provides cut resistance and a comfortable fit
  • Dexterity for improved productivity
  • Nitrile coating enhances grip in light oil applications
  • Dark coating hides dirt for longer wear

Compliance

ANSI™ Certified Level 2

Description & Specifications

Specifications

Blue,Yellow
Level 2
Nitrile
Metal fabrication, Glass handling, stamping, automotive and aerospace assembly