Kimberly-Clark Professional™ KleenGuard™ G60 Nitrile-Coated Cut-Resistant Gloves
Industrial gloves offer ANSI Level A2 Cut Protection and can withstand intermittent heat contact up to 212°F. Nitrile coating provides abrasion resistance without sacrificing dexterity or tactility.
Manufacturer:  Kimberly-Clark Professional™ KC98233
 View more versions of this product
Catalog No. 19-065-794
Description
Description
  • Cut resistant and abrasion resistant nitrile coating enhances grip capabilities
  • 15-gauge liner provides dexterity and increases performance
  • Hand-specific design ensures a snug, comfortable fit
  • Suitable for intermittent heat contact up to 212°F
Specifications
Specifications
Blue
Metal Fabrication, Glass Handling, and, Automotive Assembly
Nylon
X-Large
Nitrile
Specific
Can Withstand Intermittent Heat Contact Up to 212°F
Continuous Knit
KleenGuard™ G60
Yellow
ANSI A2
15