Kimberly-Clark Professional™ Procedure Gowns
Gowns are made with a protective fluid-resistant 3-layer SMS fabric and are recommended where there is the potential for low to moderate exposure to fluids or chemicals
Manufacturer:  Kimberly-Clark Professional™ KC69025
 View more versions of this product
Catalog No. 29-633-664
Description
Description
  • Protective three-layer SMS fabric
  • Knit cuffs
  • Velcro™ neck closure
  • Tie waist
  • Full back
Specifications
Specifications
Velcro™ neck
Blue
Large
Knit
SMS