Please login to your online account to display your discounted pricing
Kimberly-Clark™ Munising LP™ Cleanroom Papers
Ideal for the electronics, semiconductor, pharmaceutical, and medical cleanroom
$45.67 - $45.67
Products
Catalog Number Mfr. No. Price Quantity    
19-062665
kimberly clark corp
KC413127
Pack of 250 Each for $45.67
Case of 10 Pack for $400.50
Description & Specifications
Paper's antistatic properties reduce attraction of airborne particles and enhance its performance in photocopiers and converting processes.
  • Heat-stability allows it to run easily through photocopiers and laser printers
  • High opacity allows printing on both sides of the sheet without show-through
  • Latex saturated to minimize particulates and reduce the potential for particle-releasing tears
  • Can be sterilized by gamma irradiation, ethylene oxide, or steam without seriously impairing the sheet
  • Chemical- and moisture-resistant
Packaged in ISO Class 5 (formerly Class 100) cleanroom and double shrink-wrapped in packs of 250 sheets. Computer-form paper comes 1700 forms to a pack.