Please login to your online account to display your discounted pricing
Kimberly-Clark™ Professional KleenGuard™ Eyewear: V40
Comfort, flexibility, and protection
$89.98 - $89.98
Specifications
Lens Type Impact-resistant
Temple Style Dual-molded rubber
View More Specs
Products
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity    
19-167-776
kimberly-clark™ professional
08154
Uncoated Smoke Pack of 12 Each for $89.98
Case of 24 Pack for $2,007.60
Description & Specifications
Specifications
Lens Type Impact-resistant
Temple Style Dual-molded rubber
  • Impact-resistant lenses offer 99.9% UVA/UVB protection
  • Amber: For low-light environments where contrast is low; blocks out blue light to highlight and define shapes
  • Clear: For indoor applications; allows maximum visible light transmission
  • Smoke: For applications where eyestrain is a concern or bright, sunny days requiring maximum glare reduction
  • Silver Mirror: For outdoors where very bright sunlight and glare can cause eyestrain and fatigue
  • Indoor/Outdoor: For moving between indoor/shaded areas to direct sunlight and back
V40 Contour Model
  • Wraparound lens for better eye coverage
  • Soft, adjustable nose bridge
  • Comfortable, dual-molded rubber temples
  • Cushioned brow bar for additional comfort
Meet ANSI™ Z87.1-2003 standards.