Kimberly-Clark™ Professional KleenGuard™ Eyewear: V30
Four styles offer comfort, flexibility, and protection
$50.90 - $50.90
Specifications
Lens Type Impact-resistant
Temple Style Flexible
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Lens Tint Price Quantity  
Catalog Number Mfr. No. Lens Tint Price Quantity  
19-167-771 Kimberly-Clark™ Professional
KC08148
Smoke Pack of 12 Each for $50.90
Case of 24 Pack for $1,135.44
Specifications
Description
  • Impact-resistant lenses offer 99.9% UVA/UVB protection
  • Amber: For low-light environments where contrast is low; blocks out blue light to highlight and define shapes
  • Clear: For indoor applications; allows maximum visible light transmission
  • Smoke: For applications where eyestrain is a concern or bright, sunny days requiring maximum glare reduction
  • Indoor/Outdoor: For moving between indoor/shaded areas to direct sunlight and back
V30 Flexible Model
  • Wraparound lens for better eye coverage
  • Comfortable, padded nose bridge
  • Special flexible temples for added comfort
  • Adjustable lens angle
Compliance
Meet ANSI™ Z87.1-2003 standards.
Description & Specifications
Specifications
Impact-resistant
Flexible