missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ Kimguard One-Step™ KC200 Sterilization Wraps

Two sheets bonded together, making it possible to double wrap in single step

$145.04 - $382.65

Specifications

Color Light Blue
For Use With (Application) Use to wrap parts, trays and kits prior to steam, gas or gamma irradiation
Sterility Sterile
Latex Free Yes
Material SMS Fabric
View More Specs

Products 5
Catalog Number Mfr. No. Dimensions (L x W) Length (Metric) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Length (Metric) Width (Metric) Price Quantity  
06-667-01
View Documents
Kimberly-Clark Professional™
62012
12 x 12 in. (30.5 x 30.5cm) 30.5cm 30.5cm
Case of 480 Each for $145.04
 
06-667-03
View Documents
Kimberly-Clark Professional™
KC62018
18 x 18 in. (45.7 x 45.7cm) 45.7cm 45.7cm
Case of 480 Each for $318.87
 
06-667-04
View Documents
Kimberly-Clark Professional™
KC62024
24 x 24 in. (61 x 61cm) 61cm 61cm
Case of 240 Each for $294.26
 
18-892-145
View Documents
Kimberly-Clark Professional™
KC62036
36 x 36 in. (91.5 x 91.5cm) 91.5cm 91.5cm
Case of 144 Each for $382.65
 
19-065-709
View Documents
Kimberly-Clark Professional™
KC62054
54 x 54 in. (137 x 137cm) 137cm 137cm
Case of 48 Each for $309.70
 
Description

Description

KC200 is the second of six strength levels in the KIMGUARD ONE-STEP line. Use to wrap parts, trays and kits prior to steam, gas or gamma irradiation.

  • Patented design allows packages to be wrapped and opened in single step to save time
  • Imprinted with its name, size and lot code for convenience
  • Light blue SMS fabric
  • Non-sequential wrap
  • Latex free
  • Steam- and EtO-compatible
  • Recommended for lightweight packages

Specifications

Specifications

Light Blue
Sterile
SMS Fabric
Use to wrap parts, trays and kits prior to steam, gas or gamma irradiation
Yes
Steam and EtO
SDS