Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional KC100 Kimguard One-Step™ Sterilization Wraps

Two sheets bonded together, making it possible to double wrap in a single step

$161.05 - $358.51

Specifications

Color Light Blue
Design Low-lint
For Use With (Application) Light linen packs, linen gowns, towel packs, single basins and other lighter wrapping needs
Sterile No
Latex Free Yes
View More Specs
Products
Catalog Number Mfr. No. Width (English) Width (Metric) Length (English) Length (Metric) Price Quantity    

19-036-711

 
kimberly-clark™ professional
KC12730
30 in. 76.2cm 30 in. 76.2cm Case of 144 Each for $161.05

19-041-901

 
kimberly-clark™ professional
KC12718
18 in. 45.7cm 18 in. 45.7cm Case of 480 Each for $205.59

19-041-981

 
kimberly-clark™ professional
KC12724
24 in. 61cm 24 in. 61cm Case of 240 Each for $198.29

19-041-986

 
kimberly-clark™ professional
KC12748
48 in. 121.92cm 48 in. 121.92cm Case of 120 Each for $358.51
Description & Specifications

Specifications

Color Light Blue
Design Low-lint
For Use With (Application) Light linen packs, linen gowns, towel packs, single basins and other lighter wrapping needs
Sterile No
Latex Free Yes
Material SMS Fabric
Sterilization Method Steam and EtO

KC100 is the first of six strength levels in the Kimguard One-Step line and is often used for light linen packs, linen gowns, towel packs, single basins and other lighter wrapping needs.

  • Soft, strong and tear-resistant
  • Resists event-related contamination
  • One application is equivalent to two wraps
  • Naturally latex-free, nonsterile, and steam and EtO compatible
  • Fabric is imprinted with its name, size and lot code
  • Color: Light blue

Five additional strength levels (KC200-KC600) available. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information.