missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark™ Professional KC300 Kimguard™ Sterilization Wraps

Kimberly-Clark™ Professional KC300 Kimguard™ Sterilization Wraps is the third of six strengths of sterilization wrap

$202.65 - $441.37

Specifications

Color Medium Blue
Sterility Sterile
Sterilization Method Steam and EO compatible
View More Specs

Products 6
Catalog Number Mfr. No. Height (English) Height (Metric) Price Quantity  
Catalog Number Mfr. No. Height (English) Height (Metric) Price Quantity  
19-042-151 View Documents Kimberly-Clark™ Professional
KC10824
24 in. Case of 500 Each for $336.73
19-042-152 View Documents Kimberly-Clark™ Professional
KC10830
Case of 300 Each for $315.62
19-042-153 View Documents Kimberly-Clark™ Professional
KC10836
Case of 300 Each for $441.37
19-042-155 View Documents Kimberly-Clark™ Professional
KC10845
Case of 100 Each for $202.65
19-042-156 View Documents Kimberly-Clark™ Professional
KC10848
48 in. 121.92cm Case of 100 Each for $247.58
19-042-154 View Documents Kimberly-Clark™ Professional
KC10840
40 in. 101.6cm Case of 250 Each for $414.03
Description

Description

  • Kimberly-Clark has six grades of wrap fabric—from KC100 to KC600—to offer precisely the right protection
  • The higher the number, the stronger the wrap
  • Low-lint SMS fabric
  • Steam and EO compatible
  • Industry standard for sterile processing teams around the world
  • Tear-, flame-, lint- and abrasion-resistant
  • Powerguard technology for enhanced microbial barrier protection
  • Pre-vacuum steam sterilant penetration
  • Ethylene Oxide (EO) sterilant penetration and residuals
  • Maintenance of package integrity (MPI) to meet FDA requirements
  • Bacterial Filtration Efficiency (BFE) from 98.9% to 99.9%
  • Instructions for use that meet FDA guidelines
  • Class 1 flammability rating
Specifications

Specifications

Medium Blue
Sterile
Steam and EO compatible