Kimberly-Clark Professional™ KC500 Kimguard™ Sterilization Wraps
Offers precisely right protection for specific needs
Manufacturer:  Kimberly-Clark Professional™ KC68190
 View more versions of this product
Catalog No. 19-065-706
Description
Description
Offers precisely right protection for specific needs
  • Sequential wrap
  • Used for packaging of linen towels, packs, gowns, light basin sets, and single instruments for sterilization
  • Low-lint fabric enables prevention from contamination after sterilization
  • Steam and EO compatible
  • Naturally latex-free
  • Non-sterile
  • Available in multiple sizes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Order Info
Case count indicates the number of sheets included.
Specifications
Specifications
Light Blue
54 x 90 in. (137.1 x 228.6cm)
54 in.
SMS Fabric, Polypropylene
54 x 90 in. (137.1 x 228.6cm)
90 in.
Non-sterile
Low-lint
For linen towels, packs and gowns, light basin sets and single instruments and medical products
137.1cm
Yes
Steam and EtO
228.6cm