missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ KC200 Kimguard™ Sterilization Wraps

Soft, drapable wrap withstands tough handling

$137.51 - $383.21

Specifications

Color Light Blue
Design Low-lint
For Use With (Application) For linen towels, packs and gowns, light basin sets and single instruments and medical products
Latex Free Yes
Sterilization Method Steam and EtO
View More Specs

Products 3
Catalog Number Mfr. No. Length (English) Material Width (English) Price Quantity  
Catalog Number Mfr. No. Length (English) Material Width (English) Price Quantity  
06-667-35
View Documents
Kimberly-Clark Professional™
KC68012
12 in. SMS Fabric 12 in.
Case of 1000 Each for $137.51
 
19-167-795
View Documents
Kimberly-Clark Professional™
68054
54 in. SMS Fabric 54 in.
Case of 100 Each for $278.11
 
19-065-702
View Documents
Kimberly-Clark Professional™
KC68072
54 in. SMS Fabric, Polypropylene 72 in.
Case for $383.21
 
Description

Description

Second of six strength levels in the Kimguard line and is often used for linen towels, packs and gowns, light basin sets and single instruments.

  • Soft, strong and tear-resistant
  • Resists event-related contamination
  • Tough three-layer, low-lint SMS construction resists penetration by particles and liquids
  • Naturally latex-free, nonsterile, and steam and EtO compatible
  • Light blue

Order Info

Five additional strength levels (KC100, KC300, KC400, KC500 and KC600) available. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information.

Specifications

Specifications

Light Blue
For linen towels, packs and gowns, light basin sets and single instruments and medical products
Steam and EtO
Low-lint
Yes
SDS
Product Certifications