missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Kimberly-Clark Professional™ KC100 Kimguard™ Sterilization Wraps

Soft, drapable wrap withstands tough handling

$115.80 - $622.87

Specifications

For Use With (Application) Used to package linen towels, packs, gowns, light basin sets and single instruments for sterilization
Latex Free Yes
View More Specs

Products 4
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
19-029881
View Documents
Kimberly-Clark Professional™
10712
Case of 1000 Each for $115.80
 
19-029883
View Documents
Kimberly-Clark Professional™
KC10730
Case of 300 Each for $276.73
 
19-041-910
View Documents
Kimberly-Clark Professional™
KC10772
Case of 100 Each for $350.61
 
19-130-160
View Documents
Kimberly-Clark Professional™
KC10720
Case of 1000 Each for $622.87
 
Description

Description

Soft, drapable wrap withstands tough handling

First of six strength levels in the Kimguard line is often used for light linen packs and gowns, towel packs, light basins and other lighter wrapping needs.

  • Soft, strong and tear-resistant
  • Resists event-related contamination
  • Tough three-layer, low-lint SMS construction resists penetration by particles and liquids
  • Naturally latex-free, nonsterile, and steam and EtO compatible
  • Color: Light blue

Order Info

Five additional strength sequential (single sheet) wrap levels (KC200-KC600) available. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information.

Specifications

Specifications

Used to package linen towels, packs, gowns, light basin sets and single instruments for sterilization
Yes
SDS
Product Certifications