Kimberly-Clark™ Professional KC400 Microcool™ Surgical Gowns
Designed for advanced levels of protection and comfort in a surgical environment
$316.55 - $316.55
Specifications
Material Impervious Fabric
Sterile Yes
Weight (English) 18.03 lb.
Weight (Metric) 8.17kg
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-812-950 Kimberly-Clark™ Professional
KC92340
Large Case of 30 Each for $316.55
Specifications
Description
  • Enhanced ultra SMS fabric is sterile and breathable
  • Generous, book-fold cut and added length
  • Hook-and-loop adjustable neckline with orange binding
  • Raglan sleeves
Description & Specifications
Specifications
Impervious Fabric
Yes
18.03 lb.
8.17kg